2-Ethyl-3-methylpyrazine, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ300

W315508-100G-K W315508-1KG-K W315508-5KG-K
SIALW315508-100G-KEA 191 EUR
SIALW315508-100G-K SIALW315508-1KG-K SIALW315508-5KG-K
2-Ethyl-3-methylpyrazine, Sigma-Aldrich®
2-Ethyl-3-methylpyrazine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 170
Density: 0.99 g/cm³ (20 °C)
MDL Number: MFCD00006150
CAS Number: 15707-23-0
EINECS: 239-799-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR