1-Phenyl-1H-pyrazole ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14270.09
A14270.18
A14270.30
A14270.09EA
67.9
EUR
A14270.09
A14270.18
A14270.30
1-Phenyl-1H-pyrazole ≥98%
1-Phenyl-1H-pyrazole
Purity: 98%
CAS No.: 1126-00-7
Molecular Formula: C9H8N2
Formula Weight: 144.18
Flash Point: 109°(228°F)
Refractive Index: 1.5960
Boiling Point: 141-142°/30mm
Beilstein Reference: 2594
MDL No.: MFCD00003112
Formula:
C₉H₈N₂ Boiling Pt: 141…142 °C (30 mmHg) Melting Pt: 11 °C Density: 1.090 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003112 CAS Number: 1126-00-7 EINECS: 214-415-1 |