4-Methylpyridazine

Supplier: Thermo Scientific
Warning

KM08278.250 KM08278.1
MAYBKM08278.250EA 0 EUR
MAYBKM08278.250 MAYBKM08278.1
4-Methylpyridazine
4-Methylpyridazine
Formula: C₅H₆N₂
Boiling Pt: 66 °C (0,8 mmHg)
Density: 1,0641 g/cm³ (20 °C)
MDL Number: MFCD00011594
CAS Number: 1120-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR