2-Amino-5-bromopyridine

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine

BTB03304.1 BTB03304.10 BTB03304.50
MAYBBTB03304.1EA 0 EUR
MAYBBTB03304.1 MAYBBTB03304.10 MAYBBTB03304.50
2-Amino-5-bromopyridine
2-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR