2-Chloro-3-nitropyridine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
C61607-25G
C61607-5G
SIALC61607-25GEA
73.1
EUR
SIALC61607-25G
SIALC61607-5G
2-Chloro-3-nitropyridine, Sigma-Aldrich®
2-Chloro-3-nitropyridine
Formula:
C₅H₃ClN₂O₂ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006232 CAS Number: 5470-18-8 EINECS: 226-799-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...