Pyridine-2-carbonitrile, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

C94602-100G C94602-500G C94602-5G
SIALC94602-100GEA 37.7 EUR
SIALC94602-100G SIALC94602-500G SIALC94602-5G
Pyridine-2-carbonitrile, Sigma-Aldrich®
Pyridine-2-carbonitrile
Formula: C₆H₄N₂
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 26…28 °C
Density: 1.081 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006218
CAS Number: 100-70-9
EINECS: 202-880-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR