Nicotinic acid (Niacin), Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
N4126-100G N4126-1KG N4126-500G N4126-5G
SIALN4126-100GEA 56.4 EUR
SIALN4126-100G SIALN4126-1KG SIALN4126-500G SIALN4126-5G
Nicotinic acid (Niacin), Sigma-Aldrich®
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552