Isonicotinic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
M-Clarity™ quality level = MQ200
I17508-100G I17508-500G I17508-5G
SIALI17508-100GEA 29.8 EUR
SIALI17508-100G SIALI17508-500G SIALI17508-5G
Isonicotinic acid, Sigma-Aldrich®
Isonicotinic acid
Formula: C₆H₅NO₂
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 315…319 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206