Pyridine-3,4-dicarboxylic acid
Supplier: Thermo Scientific
Warning
Synonyms: 3,4-Pyridinedicarboxylic acid, Cinchomeronic acid, Pyridine-3,4-dicarboxylicacid
SB02021.1 SB02021.10 SB02021.50
MAYBSB02021.1EA 0 EUR
MAYBSB02021.1 MAYBSB02021.10 MAYBSB02021.50
Pyridine-3,4-dicarboxylic acid
Pyridine-3,4-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 529
Melting Pt: 257
Storage Temperature: Ambient
MDL Number: MFCD00006392
CAS Number: 490-11-9
EINECS: 207-705-4
Merck Index: 13,02304