Nicotinamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
72340-1KG 72340-100G 72340-250G
SIAL72340-1KGEA 225 EUR
SIAL72340-1KG SIAL72340-100G SIAL72340-250G
Nicotinamide, Sigma-Aldrich®
Nicotinamide
Formula: C₆H₆N₂O
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 127…128 °C
Density: 1.4463 g/cm³ (20 °C)
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574