Nicotinamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
M-Clarity™ quality level = MQ200
N3376-1KG N3376-500G
SIALN3376-1KGEA 102 EUR
SIALN3376-1KG SIALN3376-500G
Nicotinamide, Sigma-Aldrich®
Nicotinamide
Formula: C₆H₆N₂O
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 127…128 °C
Density: 1.4463 g/cm³ (20 °C)
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574