Isonicotinamide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Pyridinecarboxylic acid amide
M-Clarity™ quality level = MQ200
I17451-100G I17451-25G
SIALI17451-100GEA 57.8 EUR
SIALI17451-100G SIALI17451-25G
Isonicotinamide, Sigma-Aldrich®
Isonicotinamide
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Melting Pt: 155…157 °C
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2