2,6-Difluoropyridine, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

233277-25G 233277-5G
SIAL233277-25GEA 113 EUR
SIAL233277-25G SIAL233277-5G
2,6-Difluoropyridine, Sigma-Aldrich®
2,6-Difluoropyridine
Formula: C₅H₃F₂N
Boiling Pt: 123…124 °C (1013 hPa)
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006227
CAS Number: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR