2-Bromo-4-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Bromo-4-picoline
M-Clarity™ quality level = MQ100
349984-10G 349984-1G
SIAL349984-10GEA 134 EUR
SIAL349984-10G SIAL349984-1G
2-Bromo-4-methylpyridine, Sigma-Aldrich®
2-Bromo-4-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 42…44 °C (0.6 mmHg)
Melting Pt: 218…228 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00082590
CAS Number: 4926-28-7