2-Chloro-4-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Chloro-4-picoline
M-Clarity™ quality level = MQ100
116327-25ML 116327-5ML
SIAL116327-25MLEA 249 EUR
SIAL116327-25ML SIAL116327-5ML
2-Chloro-4-methylpyridine, Sigma-Aldrich®
2-Chloro-4-methylpyridine
Formula: C₆H₆ClN
Boiling Pt: 194…195 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.456 g/cm³ (25 °C)
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2