4-Acetylpyridine 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Methyl 4-pyridyl ketone
102700100 102701000 102705000
ACRO102700100EA 25.2 EUR
ACRO102700100 ACRO102701000 ACRO102705000
4-Acetylpyridine 98%
4-Acetylpyridine
Formula: C₇H₇NO
Boiling Pt: 212 °C (760 mmHg)
Melting Pt: 13…16 °C
Density: 1.09 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006433
CAS Number: 1122-54-9
EINECS: 214-350-9
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20°C, 589 nm)
GC ≥97.5 %