2-Amino-3-methylpyridine 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 97%
CAS No.: 1603-40-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 118°(244°F)
Refractive Index: 1.5825
Odour: Unpleasant
Melting Point: 29-32°
Boiling Point: 221-222°
Beilstein Reference: 107892
UN No.: 2811
MDL No.: MFCD00006320
Formula:
C₆H₈N₂ Boiling Pt: 221…222 °C Melting Pt: 29…32 °C Density: 1.073 Flash Pt: 118 °C (244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006320 CAS Number: 1603-40-3 EINECS: 216-501-4 UN: 2811 ADR: 6.1,III |