4,4'-Bipyridyl, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
289426-25G 289426-5G
SIAL289426-25GEA 148 EUR
SIAL289426-25G SIAL289426-5G
4,4'-Bipyridyl, Sigma-Aldrich®
4,4'-Bipyridyl
Formula: C₁₀H₈N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006416
CAS Number: 553-26-4
EINECS: 209-036-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,03383