5-Bromopyrimidine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
219142-25G 219142-5G
SIAL219142-25GEA 64.8 EUR
SIAL219142-25G SIAL219142-5G
5-Bromopyrimidine, Sigma-Aldrich®
5-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1