4-Methylpyrimidine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13170.06
A13170.14
A13170.06EA
73.6
EUR
A13170.06
A13170.14
4-Methylpyrimidine ≥98%
4-Methylpyrimidine
Purity: 98%
CAS No.: 3438-46-8
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 40°(104°F)
Refractive Index: 1.4960
Boiling Point: 141-142°
Beilstein Reference: 105781
UN No.: 1993
MDL No.: MFCD00006115
Formula:
C₅H₆N₂ Boiling Pt: 141…142 °C Melting Pt: 32 °C Density: 1.030 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006115 CAS Number: 3438-46-8 EINECS: 222-344-2 UN: 1993 ADR: 3,III |