2-Acetyl-1-methyl-1H-pyrrole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
M-Clarity™ quality level = MQ300
W318418-100G-K W318418-1KG-K W318418-25G-K
SIALW318418-100G-KEA 345 EUR
SIALW318418-100G-K SIALW318418-1KG-K SIALW318418-25G-K
2-Acetyl-1-methyl-1H-pyrrole, Sigma-Aldrich®
2-Acetyl-1-methyl-1H-pyrrole
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 200
Density: 1.0445 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003089
CAS Number: 932-16-1
EINECS: 213-247-6