HMDS (1,1,1,3,3,3-hexamethyldisilazane), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexamethyldisilazane
M-Clarity™ quality level = MQ200
86944-20KG-F 86944-500G-F 86944-5KG-F
SIAL86944-20KG-FEA 2740 EUR
SIAL86944-20KG-F SIAL86944-500G-F SIAL86944-5KG-F
HMDS (1,1,1,3,3,3-hexamethyldisilazane), Sigma-Aldrich®
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.7742 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 2924
ADR: 3,II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM