Hexamethylenetetramine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Hexamethylenetetramine, Hexamine, Urotropine, Formin
M-Clarity™ quality level = MQ200
H11300-1KG
H11300-25G
H11300-4KG
H11300-500G
SIALH11300-1KGEA
98.1
EUR
SIALH11300-1KG
SIALH11300-25G
SIALH11300-4KG
SIALH11300-500G
Hexamethylenetetramine, Sigma-Aldrich®
Methenamine
Formula:
C₆H₁₂N₄ Melting Pt: 280 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006895 CAS Number: 100-97-0 EINECS: 202-905-8 UN: 1328 ADR: 4.1,III Merck Index: 13,05994 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...