2,5-Dibromothiophene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
108472-100G 108472-25G
SIAL108472-100GEA 103 EUR
SIAL108472-100G SIAL108472-25G
2,5-Dibromothiophene, Sigma-Aldrich®
2,5-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 211
Melting Pt: –6 °C
Density: 2.1531 g/cm³ (13.8 °C)
MDL Number: MFCD00005420
CAS Number: 3141-27-3
EINECS: 221-547-3
UN: 2810
ADR: 6.1 II