2-Bromothiophene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Thienyl bromide

M-Clarity™ quality level = MQ100

124168-10G 124168-250G 124168-50G
SIAL124168-10GEA 39.2 EUR
SIAL124168-10G SIAL124168-250G SIAL124168-50G
2-Bromothiophene, Sigma-Aldrich®
2-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: ∼149 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.71 g/cm³ (20 °C)
Flash Pt: 52
Storage Temperature: Refrigerator
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR