3-Bromothiophene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Thienyl bromide
M-Clarity™ quality level = MQ100
106224-100G 106224-25G 106224-5G
SIAL106224-100GEA 146 EUR
SIAL106224-100G SIAL106224-25G SIAL106224-5G
3-Bromothiophene, Sigma-Aldrich®
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 158
Melting Pt: –10 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2810
ADR: 6.1 II