2-Thiophenesulphonyl chloride 98%
Supplier: Thermo Fisher Scientific
Danger
420890050 420890250
ACRO420890050EA 62.9 EUR
ACRO420890050 ACRO420890250
2-Thiophenesulphonyl chloride 98%
2-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
Boiling Pt: 80 °C (1 mmHg)
Melting Pt: 27…29 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005426
CAS Number: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to blue-green to brown-purple Low melting solid or liquid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (Ex Cl)
GC ≥97.0 %