Thiosemicarbazide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
89050-100G 89050-25G
SIAL89050-100GEA 57.3 EUR
SIAL89050-100G SIAL89050-25G
Thiosemicarbazide, Sigma-Aldrich®
Thiosemicarbazide
Formula: CH₅N₃S
Melting Pt: 178…180 °C
MDL Number: MFCD00007620
CAS Number: 79-19-6
EINECS: 201-184-7
UN: 2811
ADR: 6.1,II
Merck Index: 12,09499