4-(Chloromethyl)styrene 90% (by GC) inhibited, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Stabilization:
Inhibited with tert-butylcatechol 500 ppm
M-Clarity™ quality level = MQ200
Component of ion exchange resins, photo-resist polymers, cross-linkable fibers, coupling agents and electroconducting polymers.
Starter for various copolymer preparations. Dual functional monomer. Readily derivatised by chloride displacement.
Formula:
C₉H₉Cl Boiling Pt: 229 °C (lit.) Density: 1.083 g/lL (25 °C) (lit.) Flash Pt: 104 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051362 CAS Number: 1592-20-7 EINECS: 216-471-2 UN: 3265 ADR: 8,II |
Specification Test Results
Infrared Spectrum | Conforms to Structure |
Purity (GC) | ≥89.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...