Hydrochloric acid 1 mol/l (1N) volumetric solution
Supplier: PanReac AppliChem
Warning
Synonyms: Hydrochloric acid solution, Muriatic acid
181021.0716 181021.1211 181021.1214 181021.1315
APPC181021.0716EA 374 EUR
APPC181021.0716 APPC181021.1211 APPC181021.1214 APPC181021.1315
Hydrochloric acid 1 mol/l (1N) volumetric solution
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.011 kg/l
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III
Order Now
Specification Test Results
Titer (20 °C) 0.999 - 1.001