ESD safety shoes, lace-up, Journey S3
Supplier: ELTEN
729611/40 729611/41 729611/42 729611/43 729611/44 729611/45 729611/46 729611/47 729611/48
ELTN729611/40EA 87.9 EUR
ELTN729611/40 ELTN729611/41 ELTN729611/42 ELTN729611/43 ELTN729611/44 ELTN729611/45 ELTN729611/46 ELTN729611/47 ELTN729611/48
ESD safety shoes, lace-up, Journey S3
Shoes and Boots
Sporty ESD safety shoes made from suede, mesh and microfibre upper with reflective materials, a skin friendly textile lining, a padded tongue plus a dual-density rubber/PU sole (E-Track).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Sportive ESD)
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3