Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)
Supplier: Thermo Fisher Scientific
Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
H59536.06 H59536.14
H59536.06EA 78.9 EUR
H59536.06 H59536.14
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
Formula: C₃₈H₃₂N₂O₅
MW: 596.68 g/mol
Melting Pt: 201…204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077049
CAS Number: 132388-59-1
Order Now
Specification Test Results
Assay (HPLC) Min. 96.0%