Sodium dodecyl sulphate (SDS) ≥95% (dye basis)
Supplier: Thermo Fisher Scientific
Danger
J18220.A1 J18220.A7
J18220.A1EA 129 EUR
J18220.A1 J18220.A7
Sodium dodecyl sulphate (SDS) ≥95% (dye basis)
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: ∼206 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (Dry Basis) ≥95%
Identification (FTIR) Conforms