Starch, soluble 1% in aqueous solution for volumetric analysis, for iodometric titrations
Supplier: PanReac AppliChem
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
283146.1209 283146.1211
APPC283146.1209EA 27.1 EUR
APPC283146.1209 APPC283146.1211
Starch, soluble 1% in aqueous solution for volumetric analysis, for iodometric titrations
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
Merck Index: 13,08878