Lipopolysaccharide (from E. coli O55:B5)
Supplier: Merck
Synonyms: Lipopolysaccharide
L2880-10MG L2880-100MG L2880-25MG L2880-500MG
SAFSL2880-10MGEA 85 EUR
SAFSL2880-10MG SIALL2880-100MG SIALL2880-25MG SIALL2880-500MG
Lipopolysaccharide (from E. coli O55:B5)
Lipopolysaccharides
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria.
  • Lyophilised powder
  • Purified by phenol extraction
  • Impurities: <3% protein (Lowry)
LPS and its lipid A moiety linked to an antigenic O-polysaccharide stimulate cells of the innate immune system by the toll-like receptor 4 (TLR4), a member of the toll-like receptor protein family, which recognises common pathogen-associated molecular-patterns (PAMPs).
As part of the concanavalin A (ConA) and D-galactosamine/lipopolysaccharide (D-GalN/LPS) model of fulminant hepatitis to study the effect of the natural compound hesperetin.
Can be used to stimulate hepatocytes and non-parenchymal cells and also to stimulate the spleen and study the expression of the Malitaf gene in the fish blunt snout bream.
Packaging: 10 mg in poly bottle and 25, 100, 500 mg in glass bottle.