Culture media, in petri dishes, tryptic soy agar, with LTHThio
Supplier: MERCK LIFE SCIENCE
1467830020 1467830200
HEIP1467830020EA 74.3 EUR
HEIP1467830020 HEIP1467830200
Culture media, in petri dishes, tryptic soy agar, with LTHThio
Media for Microbiology Prepared Media for Microbiology
TSA can be used in environmental monitoring for isolators and clean rooms (surface and air monitoring). Also used for the detection of fungi and bacteria.
TSA contains CASO agar, casein soybean digest agar with lecithin, tween, histidin and thiosulphate, tryptone soya agar, tryptic soy agar with neutraliser.
Packaging: Packaged in fibre case.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions 15...25 °C