HPLC columns, NUCLEODUR® PFP
Supplier: MACHEREY-NAGEL
760431.20 760431.30 760431.40 760431.46 760433.20 760433.30 760433.40 760433.46 760435.20 760436.20 760436.30 760436.40 760436.46 760438.20 760443.20 760443.30 760443.40 760443.46 760445.46 760446.20 760446.30 760446.40 760446.46 760447.20 760447.30 760447.40 760447.46 760448.20 760448.30 760448.40 760448.46 760449.20 760449.30 760449.40 760449.46 760453.20 760453.30 760453.40 760453.46 760455.46 760456.20 760456.30 760456.40 760456.46 760457.20 760457.30 760457.40 760457.46 760458.20 760458.30 760458.40 760458.46 760459.20 760459.30 760459.40 760459.46 761975.20 761975.30 761976.20 761976.30 761977.20 761977.30
MANA760431.20EA 685 EUR
MANA760431.20 MANA760431.30 MANA760431.40 MANA760431.46 MANA760433.20 MANA760433.30 MANA760433.40 MANA760433.46 MANA760435.20 MANA760436.20 MANA760436.30 MANA760436.40 MANA760436.46 MANA760438.20 MANA760443.20 MANA760443.30 MANA760443.40 MANA760443.46 MANA760445.46 MANA760446.20 MANA760446.30 MANA760446.40 MANA760446.46 MANA760447.20 MANA760447.30 MANA760447.40 MANA760447.46 MANA760448.20 MANA760448.30 MANA760448.40 MANA760448.46 MANA760449.20 MANA760449.30 MANA760449.40 MANA760449.46 MANA760453.20 MANA760453.30 MANA760453.40 MANA760453.46 MANA760455.46 MANA760456.20 MANA760456.30 MANA760456.40 MANA760456.46 MANA760457.20 MANA760457.30 MANA760457.40 MANA760457.46 MANA760458.20 MANA760458.30 MANA760458.40 MANA760458.46 MANA760459.20 MANA760459.30 MANA760459.40 MANA760459.46 MANA761975.20 MANA761975.30 MANA761976.20 MANA761976.30 MANA761977.20 MANA761977.30
HPLC columns, NUCLEODUR® PFP
Chromatography Columns
NUCLEODUR® PFP is a pentafluorophenyl-propyl modified silica phase for HPLC.
  • High purity spherical silica gel
  • Hydrophobic, polar RP phase with additional aromatic and steric selectivity
  • Separation mechanism based on 4 retention mechanisms (polar interactions, dipole-dipole interactions, π-π interactions, hydrophobic interactions)
The separation mechanism features an alternative selectivity in comparison to classical C18 phases. Thus NUCLEODUR® PFP is well applicable for polar compounds like pharmaceuticals and phenols, halogenated compounds, aromatic and unsaturated compounds, isomers, and basic compounds - also for LC-MS detection.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).