NANOCOLOR® Photometric tube Tests, Robot
Supplier: MACHEREY-NAGEL
985681 985603 985605 985607 985619 985621 985622 985626 985629 985630 985636 985638 985657 985664 985665 985668 985673 985674 985676 985680 985683 985688 985669
MANA985681EA 95.9 EUR
MANA985681 MANA985603 MANA985605 MANA985607 MANA985619 MANA985621 MANA985622 MANA985626 MANA985629 MANA985630 MANA985636 MANA985638 MANA985657 MANA985664 MANA985665 MANA985668 MANA985673 MANA985674 MANA985676 MANA985680 MANA985683 MANA985688 MANA985669
NANOCOLOR® Photometric tube Tests, Robot
Environmental Test Kits
Suitable for automatic water analysis on the Skalar robot SP2000 series. Precise rapid tests for all types of water and wastewater samples.
  • Time and cost savings
  • Increased productivity and flexibility
  • Increased precision and accuracy
  • Fewer user errors
  • Parallelization of processes
  • Seamless documentation
Robotic system for fully automated water analysis.
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx metal: No (except: MANA985607, MANA985676, MANA985680, MANA985681)
NanOx N: No (except: MANA985683,MANA985688)