Accutase®
Supplier: Merck
Accutase is supplied as a sterile filtered solution suitable for cell culture.
- Ready to use
- Does not contain mammalian or bacterial-derived products
Used in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up. Efficient in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines, and insect cells.