Cleaner disinfectant, SURE®
Supplier: Diversey
100892312 100892313
DIVY100892312EA 167 EUR
DIVY100892312 141-1576
Cleaner disinfectant, SURE®
Cleaning Agents Disinfectants
Concentrated cleaner disinfectant for cleaning and disinfection of all surfaces, suitable for regular use.
  • Effective in all water conditions
  • Very effective against a wide range of microorganisms, bactericidal and yeasticidal activities
  • Contains plant-based ingredients from renewable sources
  • No environmental hazard classification, no safety warnings for product in use for user
SURE® cleaner disinfectant is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver professional cleaning results - while being safe for peopleMore Product Information