Sodium thiosulphate 0.05 M (0.05N) volumetric solution
Supplier: VWR Chemicals
85813.290EA 30.8 EUR
85813.290 85813.410
Sodium thiosulphate 0.05 M (0.05N) volumetric solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7