Revised B/N surrogate mix
Supplier: Restek
31887 31887.15 31887.25 31888.15 31888.25 31889.15 31889.25 31888 31889
REST31887EA 66 EUR
REST31887 REST31887.15 REST31887.25 REST31888.15 REST31888.25 REST31889.15 REST31889.25 REST31888 REST31889
Revised B/N surrogate mix
Standards Organic Standards
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).