Acrolein
Supplier: Restek
30645 30646
REST30645EA 76.5 EUR
REST30645 REST30646
Acrolein
Standards Organic Standards
Ordering information: This product has a limited shelf life. We recommend that you order only the ampoule quantity that meets your immediate needs.
Caution: NOTE: Store standards under frozen conditions (0 °C or colder). We recommend you store ampoules lying flat (horizontally) to prevent breakage. For best results, thaw the contents of the ampoule fully and shake well before opening and using the solution.