PCR plates, 96-well, skirted, FrameStar®
Supplier: 4titude Ltd.
4TI-0960/R 4TI-0960/C 4TI-0960/G 4TI-0961 4TI-0960/B 4TI-0960/P 4TI-0960/W
FTIT4TI-0960/REA 343 EUR
FTIT4TI-0960/R FTIT4TI-0960/C FTIT4TI-0960/G FTIT4TI-0961 FTIT4TI-0960/B FTIT4TI-0960/P FTIT4TI-0960/W
PCR plates, 96-well, skirted, FrameStar®
Microplates PCR Plates
The low profile wells of these plates are shorter than "standard" profile wells, which decrease the "dead space" between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled PP tubes for optimum PCR and real-time (RT-qPCR) results
  • Rigid PC frame for added mechanical stability
  • Alphanumeric grid