ESD ankle boots, lace-up, Ergo-Med 737 XP Blueline
Supplier: ATLAS - the shoe company
15336/W12 15336/W13 15336/W14 15337/W12 15337/W13 15337/W14 15338/W12 15338/W13 15338/W14 15339/W12 15339/W13 15339/W14 15340/W12 15340/W13 15340/W14 15341/W12 15341/W13 15341/W14 15347/W14 15342/W12 15342/W13 15342/W14 15343/W12 15343/W13 15343/W14 15344/W12 15344/W13 15344/W14 15345/W12 15345/W13 15345/W14 15346/W12 15346/W13 15346/W14 15347/W12 15347/W13 15348/W12 15348/W13 15348/W14
ATLA15336/W12EA 0 EUR
ATLA15336/W12 ATLA15336/W13 ATLA15336/W14 ATLA15337/W12 ATLA15337/W13 ATLA15337/W14 ATLA15338/W12 ATLA15338/W13 ATLA15338/W14 ATLA15339/W12 ATLA15339/W13 ATLA15339/W14 ATLA15340/W12 ATLA15340/W13 ATLA15340/W14 ATLA15341/W12 ATLA15341/W13 ATLA15341/W14 ATLA15347/W14 ATLA15342/W12 ATLA15342/W13 ATLA15342/W14 ATLA15343/W12 ATLA15343/W13 ATLA15343/W14 ATLA15344/W12 ATLA15344/W13 ATLA15344/W14 ATLA15345/W12 ATLA15345/W13 ATLA15345/W14 ATLA15346/W12 ATLA15346/W13 ATLA15346/W14 ATLA15347/W12 ATLA15347/W13 ATLA15348/W12 ATLA15348/W13 ATLA15348/W14
ESD ankle boots, lace-up, Ergo-Med 737 XP Blueline
Shoes and Boots
ESD safety ankle boot made from waterproof leather with aktiv-X lining and outdoor sole: 4,5 mm rough tread profile, oil, acid and petrol resistant, anti-static, heat resistant up to 170 °C.
  • Anti-static range 7,5×10⁵ - 3,5×10⁷ Ohm
  • Breathable aktiv-X lining
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Suitable for wearing with insoles
  • Protective toecap made from steel (200 J)
  • Puncture resistant midsole
Width: 12, 13, 14
Colour: Black/blue
Certifications: EN 61340 (ESD); DGUV 112-191; EN 20345; S3