Ethylene oxide
Supplier: Restek
36005 30620
REST36005EA 58 EUR
REST36005 REST30620
Ethylene oxide
Standards Organic Standards
The test for ethylene oxide is specified in many individual drug monographs of USP24/NF19. The limit test concentration is currently 10 ppm. While the specific test solution and method will vary depending on the particular drug monograph, the solution below is suitable for most tests.
Ordering information: Ethylene oxide is available in other solvents and concentrations.