RPMI-1640 medium
Supplier: Merck
R8758-500ML R8758-100ML R8758-1L R8758-24X500ML R8758-6X1L R8758-6X500ML
SIALR8758-500MLEA 32.7 EUR
SIALR8758-500ML SIALR8758-100ML SIALR8758-1L SIALR8758-24X500ML SIALR8758-6X1L SIALR8758-6X500ML
RPMI-1640 medium
Cell Culture Media
RPMI-1640 medium with L-glutamine and sodium bicarbonate is used for maintaining cell lines and serves as a medium for different cells.
  • Sterile filtered
  • Suitable for cell culture
RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.