Poly(vinylidene fluoride), M.W. ~534,000 (by GPC), powder, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
Poly(vinylidene fluoride) PVDF is a semi crystalline non-centrosymmetric polymer which exhibits piezo-, pyro- and ferroelectric properties. It is a linear polymer that shows permanent electric dipoles perpendicular to the direction of the molecular chain.
PVDF is used in nanoporous electrodes, to synthesise composites comprising of ultrafine fibers of PVDF embedded with carbon nanotubes. It can be used in sensors, actuators, for energy storage, sphygmomanometers, pressure sensors and nanogenerators, lithium ion batteries.
Formula:
(-CH₂CF₂-)n Density: 1.74 g/ml (25 °C) |
MDL Number:
MFCD00084470 CAS Number: 24937-79-9 |
Specification Test Results
Infrared Spectrum | Conforms to Structure |
Loss on Drying | ≤2.00% |
Appearance (Color) | White |
Appearance (Form) | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...