Bottles, wide neck, amber
Supplier: WEPA PUALUS AND THEWALT
33228 33231
WEPA33228EA 220 EUR
WEPA33228 WEPA33231
Bottles, wide neck, amber
Bottles Glass Bottles
Round wide-mouth bottle according to hydrolytic class III.
  • Made of glass