Cleanroom notebooks, BCR® MyNotes
Supplier: BERKSHIRE
MNBKA5CR_U MNBKA4CR_U MNBKMEMOCR MNBKA5GRID MNBKA4GRID MNBKA4CR MNBKA5CR
BERKMNBKA5CR_UEA 20.8 EUR
BERKMNBKA5CR_U BERKMNBKA4CR_U BERKMNBKMEMOCR BERKMNBKA5GRID BERKMNBKA4GRID BERKMNBKA4CR BERKMNBKA5CR
Cleanroom notebooks, BCR® MyNotes
Notebooks
BCR® MyNotes Notebooks are an ideal choice for documentation. They are practical for note taking and data recording in the cleanroom (Class 1 - 10,000).
  • Made with Berkshire cleanroom bond 850 (medium 85 g/m² (24 lbs.))
  • Low in particles and fibres
  • Full lot tracking on every case
  • Double packed for cleanliness
  • Smooth surfaces reduce particle contamination
  • Available in college rule (CR) or 1/4" grid options
BCR® Mynotes are made with the purest quality cellulose from specially selected forests and is specially coated to minimise particle and fiber release.
Certifications: Meets RoHS guidelines and designed for use in ISO Class 3-7, EU grade B/C/D.